Assay Details
| Gene Symbol: LIPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194503.2 | NP_001181432.2 | 463 | 83 | ||
| GenBank mRNA | JV060616.1 | - | 63 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIPT2| Gene Symbol: LIPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194503.2 | NP_001181432.2 | 463 | 83 | ||
| GenBank mRNA | JV060616.1 | - | 63 | 83 | ||
Set Membership: |
|