Assay Details
| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194514.3 | NP_001181443.1 | 340 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC3L| Gene Symbol: TRAPPC3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194514.3 | NP_001181443.1 | 340 | 93 | ||