Assay Details
Gene Symbol: SRXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194557.2 | NP_001181486.1 | 221 | 74 | ||
GenBank mRNA | JU323278.1 | - | 227 | 74 | ||
JU323279.1 | - | 227 | 74 | |||
JV045263.1 | - | 232 | 74 | |||
JV602539.1 | - | 247 | 74 | |||
JV704411.1 | - | 227 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
