Assay Details
| Gene Symbol: CLDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194562.2 | NP_001181491.1 | 358 | 76 | ||
| GenBank mRNA | JV044109.1 | - | 167 | 76 | ||
| JV635963.1 | - | 101 | 76 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194562.2 transcripts.
Set Membership: |
|

CLDN3
Genomic Map