Assay Details
| Gene Symbol: RFXANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194670.2 | NP_001181599.1 | 856 | 63 | ||
| XM_015123382.2 | XP_014978868.1 | 917 | 63 | |||
| XM_015123383.2 | XP_014978869.1 | 911 | 63 | |||
| XM_015123384.2 | XP_014978870.1 | 918 | 63 | |||
| XM_028838541.1 | XP_028694374.1 | 925 | 63 | |||
| XM_028838542.1 | XP_028694375.1 | 750 | 63 | |||
| XM_028838543.1 | XP_028694376.1 | 919 | 63 | |||
| XM_028838544.1 | XP_028694377.1 | 921 | 63 | |||
| GenBank mRNA | JU336876.1 | - | 587 | 63 | ||
| JU477613.1 | - | 599 | 63 | |||
| JV048549.1 | - | 597 | 63 | |||
| JV566930.1 | - | 597 | 63 | |||
| JV664271.1 | - | 619 | 63 | |||
More Information
Set Membership: |
|

RFXANK
Genomic Map