Assay Details
| Gene Symbol: DNLZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194735.1 | NP_001181664.1 | 369 | 74 | ||
| XM_028834402.1 | XP_028690235.1 | 961 | 74 | |||
| GenBank mRNA | JU336199.1 | - | 391 | 74 | ||
| JU477042.1 | - | 386 | 74 | |||
| JV660357.1 | - | 399 | 74 | |||
| JV695163.1 | - | 348 | 74 | |||
More Information
Set Membership: |
|

DNLZ
Genomic Map