Assay Details
| Gene Symbol: CLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194783.1 | NP_001181712.1 | 121 | 75 | ||
| XM_015114120.2 | XP_014969606.1 | 968 | 75 | |||
| GenBank mRNA | JU319951.1 | - | 107 | 75 | ||
| JU474101.1 | - | 90 | 75 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194783.1, XM_015114120.2 transcripts.
Set Membership: |
|

CLP1
Genomic Map