Assay Details
| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194857.1 | NP_001181786.1 | 370 | 74 | ||
| XM_015133122.2 | XP_014988608.1 | 484 | 74 | |||
| XM_028844733.1 | XP_028700566.1 | 492 | 74 | |||
| XM_028844734.1 | XP_028700567.1 | 488 | 74 | |||
| GenBank mRNA | JU337009.1 | - | 437 | 74 | ||
| JV048635.1 | - | 436 | 74 | |||
| JV597846.1 | - | 436 | 74 | |||
More Information
Set Membership: |
|

ZFAND2A
Genomic Map