Assay Details
| Gene Symbol: PLA2G12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194877.3 | NP_001181806.1 | 469 | 94 | ||
| GenBank mRNA | JU321069.1 | - | 287 | 94 | ||
| JU332210.1 | - | 287 | 94 | |||
| JU332211.1 | - | 287 | 94 | |||
| JU332212.1 | - | 287 | 94 | |||
| JU473084.1 | - | 237 | 94 | |||
| JV636633.1 | - | 389 | 94 | |||
| Gene Symbol: DCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV046313.1 | - | 770 | 94 | ||
More Information
Set Membership: |
|

PLA2G12A
Genomic Map