Assay Details
| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190894.2 | NP_001177823.1 | 249 | 73 | ||
| XM_015137240.2 | XP_014992726.1 | 362 | 73 | |||
| GenBank mRNA | CO774134.1 | - | 253 | 73 | ||
| JV043431.1 | - | 356 | 73 | |||
| JV636222.1 | - | 355 | 73 | |||
More Information
Set Membership: |
|

PLN
Genomic Map