Assay Details
| Gene Symbol: DNASE1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194919.1 | NP_001181848.1 | 494 | 78 | ||
| XM_015128460.2 | XP_014983946.1 | 1053 | 78 | |||
| XM_015128461.2 | XP_014983947.1 | 1118 | 78 | |||
| GenBank mRNA | JU474553.1 | - | 406 | 78 | ||
More Information
Set Membership: |
|

DNASE1L1
Genomic Map