Assay Details
| Gene Symbol: MTRNR2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195028.2 | NP_001181957.1 | 346 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195028.2 transcripts.
Set Membership: |
|

MTRNR2L3
Genomic Map