Assay Details
| Gene Symbol: CCNI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195445.1 | NP_001182374.1 | 308 | 72 | ||
| XM_015138439.2 | XP_014993925.1 | 675 | 72 | |||
| GenBank mRNA | CN641931.1 | - | 308 | 72 | ||
| JU322224.1 | - | 589 | 72 | |||
| JU322226.1 | - | 589 | 72 | |||
| JU471183.1 | - | 593 | 72 | |||
| JV043690.1 | - | 601 | 72 | |||
| JV565460.1 | - | 659 | 72 | |||
More Information
Set Membership: |
|

CCNI
Genomic Map