Assay Details
| Gene Symbol: AKR1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195574.1 | NP_001182503.1 | 939 | 88 | ||
| GenBank mRNA | JU333193.1 | - | 962 | 88 | ||
| JV046318.1 | - | 968 | 88 | |||
| JV489092.1 | - | 962 | 88 | |||
| JV490552.1 | - | 962 | 88 | |||
| JV601338.1 | - | 964 | 88 | |||
| JV669510.1 | - | 1002 | 88 | |||
More Information
Set Membership: |
|

AKR1C1
Genomic Map