Assay Details
| Gene Symbol: BHLHB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193721.3 | NP_001180650.1 | 136 | 100 | ||
| XM_015127863.2 | XP_014983349.1 | 182 | 100 | |||
| XM_015127864.2 | XP_014983350.1 | 208 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193721.3, XM_015127863.2, XM_015127864.2 transcripts.
Set Membership: |
|

BHLHB9
Genomic Map