Assay Details
Gene Symbol: C3H7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193747.2 | NP_001180676.1 | 359 | 103 | ||
XM_015133126.2 | XP_014988612.1 | 464 | 103 | |||
GenBank mRNA | JV560246.1 | - | 254 | 103 |

More Information
Set Membership: |
![]() ![]() |
