Assay Details
| Gene Symbol: RAB40AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198704.1 | NP_001185633.1 | 952 | 79 | ||
| XM_028841662.1 | XP_028697495.1 | 2665 | 79 | |||
More Information
Set Membership: |
|

RAB40AL
Genomic Map