Assay Details
| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202444.1 | NP_001189373.1 | 664 | 62 | ||
| XM_015114707.2 | XP_014970193.2 | 684 | 62 | |||
| XM_028832536.1 | XP_028688369.1 | 691 | 62 | |||
More Information
Set Membership: |
|

NCR3LG1
Genomic Map