Assay Details
| Gene Symbol: NME2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204518.1 | NP_001191447.1 | 185 | 58 | ||
| XM_015119231.2 | XP_014974717.1 | 350 | 58 | |||
| XM_028835441.1 | XP_028691274.1 | 226 | 58 | |||
| GenBank mRNA | CO581875.1 | - | 189 | 58 | ||
| JV600817.1 | - | 354 | 58 | |||
More Information
Set Membership: |
|

NME2
Genomic Map