Assay Details
| Gene Symbol: BORCS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205198.1 | NP_001192127.1 | 511 | 65 | ||
| GenBank mRNA | JU320878.1 | - | 113 | 65 | ||
| JV651729.1 | - | 134 | 65 | |||
More Information
Set Membership: |
|

BORCS8
Genomic Map