Assay Details
| Gene Symbol: ICAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195749.1 | NP_001182678.1 | 934 | 98 | ||
| XM_028838471.1 | XP_028694304.1 | 1092 | 98 | |||
| GenBank mRNA | AF340063.1 | - | 897 | 98 | ||
| CO645507.1 | - | 584 | 98 | |||
| JU473373.1 | - | 981 | 98 | |||

ICAM3
Genomic Map