Assay Details
| Gene Symbol: ICAM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195749.1 | NP_001182678.1 | 1437 | 100 | ||
| XM_028838471.1 | XP_028694304.1 | 1595 | 100 | |||
| GenBank mRNA | AF340063.1 | - | 1400 | 100 | ||
| JU473373.1 | - | 1484 | 100 | |||
| JV650071.1 | - | 419 | 100 | |||
More Information
Set Membership: |
|

ICAM3
Genomic Map