Assay Details
| Gene Symbol: NME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204515.1 | NP_001191444.1 | 181 | 92 | ||
| XM_015119229.2 | XP_014974715.1 | 260 | 92 | |||
| GenBank mRNA | CN802856.1 | - | 181 | 92 | ||
| JV685161.1 | - | 142 | 92 | |||
More Information
Set Membership: |
|

NME1
Genomic Map