Assay Details
| Gene Symbol: APOBEC3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345935.1 | NP_001332864.1 | 214 | 78 | ||
| XM_015150343.2 | XP_015005829.2 | 270 | 78 | |||
| XM_028827468.1 | XP_028683301.1 | 255 | 78 | |||
| XM_028827469.1 | XP_028683302.1 | 342 | 78 | |||
| GenBank mRNA | DQ507277.1 | - | 151 | 78 | ||
| JU481501.1 | - | 214 | 78 | |||
| JU481502.1 | - | 214 | 78 | |||
| JV637136.1 | - | 259 | 78 | |||
More Information
Set Membership: |
|

APOBEC3H
Genomic Map