Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194052.2 | NP_001180981.1 | 335 | 81 | ||
| XM_001096170.4 | XP_001096170.1 | 979 | 81 | |||
| GenBank mRNA | JU336043.1 | - | 435 | 81 | ||
| JU336044.1 | - | 435 | 81 | |||
| JU336045.1 | - | 435 | 81 | |||
| JU336046.1 | - | 435 | 81 | |||
| JU336047.1 | - | 435 | 81 | |||
| JU476908.1 | - | 550 | 81 | |||
| JV043863.1 | - | 759 | 81 | |||
| JV638228.1 | - | 770 | 81 | |||
More Information
Set Membership: |
|

SNRPD3
Genomic Map