Assay Details
| Gene Symbol: NR2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194824.3 | NP_001181753.2 | 294 | 76 | ||
| XM_001114305.4 | XP_001114305.3 | 1090 | 76 | |||
| GenBank mRNA | JV574193.1 | - | 500 | 76 | ||
More Information
Set Membership: |
|

NR2F6
Genomic Map