Assay Details
| Gene Symbol: PSENEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194826.1 | NP_001181755.1 | 95 | 98 | ||
| XM_028838313.1 | XP_028694146.1 | 101 | 98 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194826.1, XM_028838313.1 transcripts.
Set Membership: |
|

PSENEN
Genomic Map