Assay Details
| Gene Symbol: TPPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194843.1 | NP_001181772.1 | 346 | 95 | ||
| XM_028840335.1 | XP_028696168.1 | 330 | 95 | |||
| GenBank mRNA | JV600761.1 | - | 318 | 95 | ||
| JV668979.1 | - | 296 | 95 | |||
More Information
Set Membership: |
|

TPPP3
Genomic Map