Assay Details
| Gene Symbol: BEX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194916.1 | NP_001181845.1 | 97 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194916.1 transcripts.
Set Membership: |
|

BEX3
Genomic Map