Assay Details
| Gene Symbol: FMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194423.2 | NP_001181352.1 | 145 | 103 | ||
| GenBank mRNA | JU323284.1 | - | 244 | 103 | ||
| JU474460.1 | - | 131 | 103 | |||
| JV674336.1 | - | 131 | 103 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194423.2 transcripts.
Set Membership: |
|

FMC1
Genomic Map