Assay Details
| Gene Symbol: ORAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194521.2 | NP_001181450.1 | 1052 | 73 | ||
| XM_028840566.1 | XP_028696399.1 | 881 | 73 | |||
| XM_028840567.1 | XP_028696400.1 | 896 | 73 | |||
| GenBank mRNA | JV045179.1 | - | 899 | 73 | ||
| JV635855.1 | - | 1045 | 73 | |||
More Information
Set Membership: |
|

ORAI3
Genomic Map