Assay Details
Gene Symbol: VDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145274.2 | XP_015000760.1 | 944 | 82 | ||
XM_015145275.2 | XP_015000761.1 | 1246 | 82 | |||
XM_015145276.2 | XP_015000762.1 | 867 | 82 | |||
XM_015145277.1 | XP_015000763.1 | 899 | 82 | |||
XM_015145278.2 | XP_015000764.1 | 799 | 82 | |||
XM_015145280.2 | XP_015000766.1 | 985 | 82 | |||
XM_015145281.2 | XP_015000767.1 | 747 | 82 | |||
XM_028852635.1 | XP_028708468.1 | 1236 | 82 | |||
XM_028852636.1 | XP_028708469.1 | 890 | 82 | |||
GenBank mRNA | JU319827.1 | - | 817 | 82 | ||
JU470804.1 | - | 807 | 82 | |||
JV044798.1 | - | 785 | 82 | |||
JV453250.1 | - | 826 | 82 | |||
JV528506.1 | - | 826 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
