Assay Details
| Gene Symbol: C7H14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190873.1 | NP_001177802.1 | 121 | 62 | ||
| GenBank mRNA | CB312361.1 | - | 124 | 62 | ||
| JU335202.1 | - | 139 | 62 | |||
| JU476179.1 | - | 132 | 62 | |||
| JV654869.1 | - | 144 | 62 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190873.1 transcripts.
Set Membership: |
|

C7H14orf119
Genomic Map