Assay Details
Gene Symbol: ZNF581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194834.2 | NP_001181763.1 | 121 | 97 |
Gene Symbol: ZNF580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839064.1 | XP_028694897.1 | 150 | 97 | ||
GenBank mRNA | JU321427.1 | - | 119 | 97 | ||
JV685926.1 | - | 137 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194834.2, XM_028839064.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
