Assay Details
| Gene Symbol: GSTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195776.1 | NP_001182705.1 | 420 | 94 | ||
| GenBank mRNA | CO582862.1 | - | 424 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSTA1| Gene Symbol: GSTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195776.1 | NP_001182705.1 | 420 | 94 | ||
| GenBank mRNA | CO582862.1 | - | 424 | 94 | ||
Set Membership: |
|