Assay Details
| Gene Symbol: MPHOSPH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193603.1 | NP_001180532.1 | 164 | 85 | ||
| XM_028840580.1 | XP_028696413.1 | 228 | 85 | |||
| GenBank mRNA | CN647132.1 | - | 59 | 85 | ||
| JU332445.1 | - | 196 | 85 | |||
| JU473911.1 | - | 183 | 85 | |||
| JV045744.1 | - | 183 | 85 | |||
More Information
Set Membership: |
|

MPHOSPH6
Genomic Map