Assay Details
| Gene Symbol: EGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258086.1 | NP_001245015.1 | 506 | 71 | ||
| GenBank mRNA | JU471706.1 | - | 506 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EGR2| Gene Symbol: EGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258086.1 | NP_001245015.1 | 506 | 71 | ||
| GenBank mRNA | JU471706.1 | - | 506 | 71 | ||
Set Membership: |
|