Assay Details
Gene Symbol: KLHL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258115.1 | NP_001245044.1 | 1948 | 85 | ||
XM_015142932.2 | XP_014998418.2 | 2292 | 85 | |||
XM_015142933.2 | XP_014998419.2 | 5502 | 85 | |||
XM_028850215.1 | XP_028706048.1 | 5199 | 85 | |||
GenBank mRNA | JU471650.1 | - | 1948 | 85 | ||
JU471651.1 | - | 1948 | 85 | |||
JU471652.1 | - | 1948 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
