Assay Details
| Gene Symbol: LITAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258181.1 | NP_001245110.1 | 249 | 78 | ||
| GenBank mRNA | JU323150.1 | - | 289 | 78 | ||
| JU472770.1 | - | 249 | 78 | |||
| JV704028.1 | - | 292 | 78 | |||
More Information
Set Membership: |
|

LITAF
Genomic Map