Assay Details
| Gene Symbol: CHST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257509.1 | NP_001244438.1 | 1419 | 73 | ||
| XM_015131917.2 | XP_014987403.1 | 2993 | 73 | |||
| GenBank mRNA | JU337176.1 | - | 1419 | 73 | ||
| JU477902.1 | - | 2044 | 73 | |||
| JV601266.1 | - | 2211 | 73 | |||
| JV727282.1 | - | 2041 | 73 | |||
More Information
Set Membership: |
|

CHST2
Genomic Map