Assay Details
| Gene Symbol: TRMT10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257537.1 | NP_001244466.1 | 736 | 74 | ||
| XM_015117398.2 | XP_014972884.2 | 1140 | 74 | |||
| XM_028834393.1 | XP_028690226.1 | 1140 | 74 | |||
| GenBank mRNA | JU323009.1 | - | 736 | 74 | ||
| JU474468.1 | - | 730 | 74 | |||
| JU474469.1 | - | 730 | 74 | |||
| JU474470.1 | - | 730 | 74 | |||
| JU474471.1 | - | 730 | 74 | |||
| JU474472.1 | - | 730 | 74 | |||
| JU474473.1 | - | 730 | 74 | |||
| JU474474.1 | - | 730 | 74 | |||
| JV044874.1 | - | 720 | 74 | |||
More Information
Set Membership: |
|

TRMT10B
Genomic Map