Assay Details
| Gene Symbol: HIGD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257901.1 | NP_001244830.1 | 195 | 70 | ||
| GenBank mRNA | JU333272.1 | - | 195 | 70 | ||
| JU474585.1 | - | 201 | 70 | |||
More Information
Set Membership: |
|

HIGD2A
Genomic Map