Assay Details
| Gene Symbol: MAGEH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260625.1 | NP_001247554.1 | 824 | 76 | ||
| GenBank mRNA | JU319598.1 | - | 811 | 76 | ||
| JU471213.1 | - | 815 | 76 | |||
| JV044954.1 | - | 824 | 76 | |||
More Information
Set Membership: |
|

MAGEH1
Genomic Map