Assay Details
| Gene Symbol: PCMTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260721.1 | NP_001247650.1 | 717 | 62 | ||
| XM_028827114.1 | XP_028682947.1 | 779 | 62 | |||
| XM_028827115.1 | XP_028682948.1 | 423 | 62 | |||
| GenBank mRNA | AY680515.1 | - | 88 | 62 | ||
| JU335287.1 | - | 717 | 62 | |||
| JU476244.1 | - | 688 | 62 | |||
| JV047534.1 | - | 717 | 62 | |||
| JV586293.1 | - | 607 | 62 | |||
More Information
Set Membership: |
|

PCMTD2
Genomic Map