Assay Details
| Gene Symbol: PLAGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260788.1 | NP_001247717.1 | 582 | 88 | ||
| XM_015137178.2 | XP_014992664.1 | 1324 | 88 | |||
| XM_015137179.2 | XP_014992665.1 | 1397 | 88 | |||
| XM_015137180.2 | XP_014992666.1 | 1365 | 88 | |||
| XM_015137183.2 | XP_014992669.1 | 1411 | 88 | |||
| XM_015137186.2 | XP_014992672.1 | 1357 | 88 | |||
| XM_015137187.2 | XP_014992673.1 | 1284 | 88 | |||
| XM_015137188.2 | XP_014992674.1 | 1338 | 88 | |||
| XM_015137189.2 | XP_014992675.1 | 1325 | 88 | |||
| XM_015137190.2 | XP_014992676.1 | 1379 | 88 | |||
| XM_015137191.2 | XP_014992677.1 | 1371 | 88 | |||
| XM_015137192.2 | XP_014992678.1 | 1339 | 88 | |||
| XM_028846682.1 | XP_028702515.1 | 9692 | 88 | |||
| XM_028846683.1 | XP_028702516.1 | 9591 | 88 | |||
| XM_028846684.1 | XP_028702517.1 | 7021 | 88 | |||
| XM_028846685.1 | XP_028702518.1 | 6989 | 88 | |||
| XM_028846686.1 | XP_028702519.1 | 5566 | 88 | |||
| GenBank mRNA | JU471235.1 | - | 842 | 88 | ||
| JV044429.1 | - | 582 | 88 | |||
| JV636219.1 | - | 739 | 88 | |||
More Information
Set Membership: |
|

PLAGL1
Genomic Map