Assay Details
| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260884.1 | NP_001247813.1 | 487 | 96 | ||
| GenBank mRNA | JU334947.1 | - | 512 | 96 | ||
| JU475951.1 | - | 520 | 96 | |||
| JV047340.1 | - | 487 | 96 | |||
More Information
Set Membership: |
|

SF3B6
Genomic Map