Assay Details
| Gene Symbol: TMEM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261045.1 | NP_001247974.1 | 96 | 107 | ||
| GenBank mRNA | JU327635.1 | - | 87 | 107 | ||
| JV045816.1 | - | 96 | 107 | |||
| JV568204.1 | - | 96 | 107 | |||
More Information
Set Membership: |
|

TMEM170B
Genomic Map