Assay Details
| Gene Symbol: SLC39A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261144.1 | NP_001248073.1 | 749 | 72 | ||
| GenBank mRNA | JU322705.1 | - | 749 | 72 | ||
| JU336367.1 | - | 505 | 72 | |||
| JU471465.1 | - | 837 | 72 | |||
| JU477190.1 | - | 531 | 72 | |||
| JV044572.1 | - | 749 | 72 | |||
| JV600319.1 | - | 500 | 72 | |||
| JV637804.1 | - | 829 | 72 | |||
More Information
Set Membership: |
|

SLC39A7
Genomic Map