Assay Details
| Gene Symbol: MALSU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261201.2 | NP_001248130.1 | 450 | 90 | ||
| GenBank mRNA | JU332817.1 | - | 445 | 90 | ||
| JU474229.1 | - | 450 | 90 | |||
| JV046123.1 | - | 439 | 90 | |||

MALSU1
Genomic Map