Assay Details
| Gene Symbol: NUDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261225.1 | NP_001248154.1 | 192 | 63 | ||
| XM_028845013.1 | XP_028700846.1 | 416 | 63 | |||
| GenBank mRNA | JU322807.1 | - | 192 | 63 | ||
| JU478156.1 | - | 204 | 63 | |||
More Information
Set Membership: |
|

NUDT1
Genomic Map